Merck
  • When do cancer patients regret their treatment decision? A path analysis of the influence of clinicians' communication styles and the match of decision-making styles on decision regret.
When do cancer patients regret their treatment decision? A path analysis of the influence of clinicians' communication styles and the match of decision-making styles on decision regret.
Patient education and counseling (2015-12-15)
Jennifer Nicolai, Angela Buchholz, Nathalie Seefried, Katrin Reuter, Martin Härter, Wolfgang Eich, Christiane Bieber
ABSTRACT
To test the influence of physician empathy (PE), shared decision making (SDM), and the match between patients' preferred and perceived decision-making styles on patients' decision regret. Patients with breast or colon cancer (n=71) completed questionnaires immediately following (T1) and three months after a consultation (T2). Path analysis was used to examine the relationships among patient demographics, patient reports of PE, SDM, the match between preferred and perceived decision-making styles, and patient decision regret at T2. After controlling for clinician clusters, higher PE was directly associated with more SDM (β=0.43, p<0.01) and lower decision regret (β=-0.28, p<0.01). The match between patients' preferred and perceived roles was negatively associated with decision regret (β=-0.33, p<0.01). Patients who participated less than desired reported more decision regret at T2. There was no significant association between SDM and decision regret (β=0.03, p=0.74). PE and the match between patients' preferred and perceived roles in medical decision making are essential for patient-centered cancer consultations and treatment decisions. Ways to enhance PE and matching the consultation style to patients' expectations should be encouraged.
Sigma-Aldrich
3-(1-Pyridinio)-1-propanesulfonate, ≥97.0% (N)