Merck
  • Palladium-catalyzed cross-coupling between vinyl halides and tert-butyl carbazate: first general synthesis of the unusual N-Boc-N-alkenylhydrazines.
Palladium-catalyzed cross-coupling between vinyl halides and tert-butyl carbazate: first general synthesis of the unusual N-Boc-N-alkenylhydrazines.
Organic letters (2007-01-16)
José Barluenga, Patricia Moriel, Fernando Aznar, Carlos Valdés
ABSTRACT
N-Boc-N-alkenylhydrazines, an almost unknown type of compounds, have been prepared with high to moderate yields via palladium-catalyzed cross-coupling between alkenyl halides and tert-butyl carbazate. The present methodology represents the first general way to access this highly functionalized and unusual type of hydrazines. [reaction: see text].
Sigma-Aldrich
tert-Butyl carbazate, 98%