- Probing the aglycon binding site of a beta-glucosidase: a collection of C-1-modified 2,5-dideoxy-2,5-imino-D-mannitol derivatives and their structure-activity relationships as competitive inhibitors.
Probing the aglycon binding site of a beta-glucosidase: a collection of C-1-modified 2,5-dideoxy-2,5-imino-D-mannitol derivatives and their structure-activity relationships as competitive inhibitors.
Bioorganic & medicinal chemistry (2004-06-10)
Tanja M Wrodnigg, Frederik Diness, Christoph Gruber, Herwig Häusler, Inge Lundt, Karen Rupitz, Andreas J Steiner, Arnold E Stütz, Chris A Tarling, Stephen G Withers, Heidrun Wölfler
PMID15186833
ABSTRACT
A range of new C-1 modified derivatives of the powerful glucosidase inhibitor 2,5-dideoxy-2,5-imino-D-mannitol has been synthesised and their biological activities probed with the beta-glucosidase from Agrobacterium sp. Ki values are compared with those of previously prepared close relatives. Findings suggest dramatic effects exerted by the aglycon binding site on substrate/inhibitor binding.