Merck
Production and radioprotective effects of pyrroloquinoline quinone.
International journal of molecular sciences (2012-01-25)
Xiang-Hua Xiong, Yan Zhao, Xin Ge, Shou-Jun Yuan, Jian-Hua Wang, Jing-Juan Zhi, Yan-Xin Yang, Bao-Hua Du, Wan-Jun Guo, Shan-Shan Wang, De-Xuan Yang, Wei-Cai Zhang
ABSTRACT
Pyrroloquinoline quinone (PQQ) was produced by fermentation of the Methylovorus sp. MP688 strain and purified by ion-exchange chromatography, crystallization and recrystallization. The yield of PQQ reached approximately 125 mg/L and highly pure PQQ was obtained. To determine the optimum dose of PQQ for radioprotection, three doses (2 mg/kg, 4 mg/kg, 8 mg/kg) of PQQ were orally administrated to the experimental animals subjected to a lethal dose of 8.0 Gy in survival test. Survival of mice in the irradiation + PQQ (4 mg/kg) group was found to be significantly higher in comparison with the irradiation and irradiation + nilestriol (10 mg/kg) groups. The numbers of hematocytes and bone marrow cells were measured for 21 days after sublethal 4 Gy gamma-ray irradiation with per os of 4 mg/kg of PQQ. The recovery of white blood cells, reticulocytes and bone marrow cells in the irradiation + PQQ group was faster than that in the irradiation group. Furthermore, the recovery of bone marrow cell in the irradiation + PQQ group was superior to that in irradiation + nilestriol group. Our results clearly indicate favourable effects on survival under higher lethal radiation doses and the ability of pyrroloquinoline quinine to enhance haemopoietic recovery after sublethal radiation exposure.
Sigma-Aldrich
Methoxatin disodium salt, ≥97.0% (HPLC)