Merck
  • Analysis of RNA-protein complexes by RNA coimmunoprecipitation and RT-PCR analysis from Caenorhabditis elegans.
Analysis of RNA-protein complexes by RNA coimmunoprecipitation and RT-PCR analysis from Caenorhabditis elegans.
Cold Spring Harbor protocols (2010-02-12)
Britta Jedamzik, Christian R Eckmann
ABSTRACT
RNA coimmunoprecipitation (co-IP) experiments are an extension of protein co-IP experiments in which in vivo RNA-protein complexes are investigated. This protocol describes how to perform RNA co-IPs from C. elegans whole-worm extracts. In principle, a protein-specific antibody is used to purify the protein of choice and its associated complex members from worm extract. This may also include RNA molecules associated with other protein components. To identify a specific mRNA molecule, all RNA molecules are first separated from the protein components after immunopurification. The mRNAs are then converted into cDNA by reverse transcription. Candidate mRNAs are detected by sensitive gene-specific amplification via polymerase chain reaction (PCR) in a semiquantitative manner. Since RNA molecules are very prone to degradation, it is crucial to avoid any kind of contamination with RNase activity in this experiment.
Product Description
Roche
RNA from yeast, dry powder, pkg of 100 g