Merck
  • Aberrant splicing of Hugl-1 is associated with hepatocellular carcinoma progression.
Aberrant splicing of Hugl-1 is associated with hepatocellular carcinoma progression.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-05-19)
Xuefeng Lu, Xiujing Feng, Xiaobo Man, Guang Yang, Liang Tang, Dan Du, Fan Zhang, Haixin Yuan, Qin Huang, Zhe Zhang, Yinkun Liu, Dennis Strand, Zhengjun Chen
ABSTRACT
Lethal giant larvae functions as a cell polarity regulator and a tumor suppressor in Drosophila. Its evolutionary conservation implies a tumor suppressor role for its human homologue, Hugl-1. The aims of this study were to characterize Hugl-1 and to determine the clinical significance of Hugl-1 alterations in hepatocellular carcinoma (HCC). Sequence alterations of Hugl-1 from 80 HCC specimens and 5 HCC cell lines were characterized by reverse transcription-PCR and sequence analysis. Western blot was used for determining Hugl-1 expression. The biological activities of Hugl-1 and its aberrant variants were examined by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay, wound healing assay, Boyden chamber assay, and tumorigenicity assay. In 32.5% (26 of 80) of the specimens and 20.0% (one of five) of HCC cell lines, 23 unique aberrant Hugl-1 transcripts were identified, most of which resulted from skipping part of and/or entire exon or insertion of intron sequences. The majority of these aberrant Hugl-1 transcripts encoded truncated proteins lacking one or more conserved WD-40 repeat motifs. Two truncated Hugl-1 proteins were found exclusively in HCC tissues. Aberrant Hugl-1 transcripts (78.3%, 20 of 23) had a short "direct repeat" sequence flanking their deleted regions. The abnormal Hugl-1 was significantly correlated with poor differentiation and large tumor size of HCC. Overexpression of two representative HCC-derived aberrant Hugl-1 variants promoted HCC cell migration, invasion, and tumorigenicity in nude mice. We provide the first evidence that Hugl-1 mRNA is frequently mutated by aberrant splicing exclusively in HCC, which may be involved in HCC progression.