Merck
  • Getting PrEPared for HIV Prevention Navigation: Young Black Gay Men Talk About HIV Prevention in the Biomedical Era.
Getting PrEPared for HIV Prevention Navigation: Young Black Gay Men Talk About HIV Prevention in the Biomedical Era.
AIDS patient care and STDs (2015-06-30)
Matt G Mutchler, Bryce McDavitt, Mansur A Ghani, Kelsey Nogg, Terrell J A Winder, Juliana K Soto
ABSTRACT
Biomedical HIV prevention strategies, such as pre-exposure prophylaxis (PrEP) and post-exposure prophylaxis (PEP), represent new opportunities to reduce critically high HIV infection rates among young black men who have sex with men (YBMSM). We report results of 24 dyadic qualitative interviews (N=48), conducted in Los Angeles, CA, exploring how YBMSM and their friends view PrEP and PEP. Interviews were analyzed using a grounded theory approach. Participants had widely divergent levels of knowledge about these prevention methods. Misconceptions and mistrust regarding PrEP were common, and concerns were expressed about PrEP-related stigma and the potential for gossip among peers who might assume a person on PrEP was HIV-positive. Yet participants also framed PrEP and PEP as valuable new options within an expanded "tool kit" of HIV prevention strategies that created possibilities for preventing new HIV infections, dating men with a different HIV status, and decreased anxiety about exposure to HIV. We organized themes around four main areas: (1) information and misinformation about biomedical HIV prevention; (2) expectations about PrEP, sexual behavior, and stigma; (3) gossip, disclosure, and "spreading the word" about PrEP and PEP; and (4) the roles of PrEP and PEP in an expanded HIV prevention tool kit. The findings suggest a need for guidance in navigating the increasingly complex array of HIV-prevention options available to YBMSM. Such "prevention navigation" could counter misconceptions and address barriers, such as stigma and mistrust, while helping YBMSM make informed selections from among expanded HIV prevention options.
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)