Merck
  • Severe cardiac phenotype with right ventricular predominance in a large cohort of patients with a single missense mutation in the DES gene.
Severe cardiac phenotype with right ventricular predominance in a large cohort of patients with a single missense mutation in the DES gene.
Heart rhythm (2009-11-03)
J Peter van Tintelen, Isabelle C Van Gelder, Angeliki Asimaki, Albert J H Suurmeijer, Ans C P Wiesfeld, Jan D H Jongbloed, Arthur van den Wijngaard, Jan B M Kuks, Karin Y van Spaendonck-Zwarts, Nicolette Notermans, Ludolf Boven, Freek van den Heuvel, Hermine E Veenstra-Knol, Jeffrey E Saffitz, Robert M W Hofstra, Maarten P van den Berg
ABSTRACT
Desmin-related myopathy is a clinically heterogenous group of disorders encompassing myopathies, cardiomyopathies, conduction disease, and combinations of these disorders. Mutations in the gene encoding desmin (DES), a major intermediate filament protein, can underlie this phenotype. The purpose of this study was to investigate the clinical and pathologic characteristics of 27 patients from five families with an identical mutation in the head domain region (p.S13F) of desmin. All 27 carriers or obligate carriers of a p.S13F DES founder mutation demonstrated a fully penetrant yet variable phenotype. All patients demonstrated cardiac involvement characterized by high-grade AV block at young ages and important right ventricular (RV) involvement. RV predominance was demonstrated by the presence of right bundle branch block in 10 patients (sometimes as a first manifestation) and by RV heart failure in 6 patients, including 2 patients who fulfilled the diagnostic criteria for arrhythmogenic RV cardiomyopathy. Because of this clinical overlap with desmosome cardiomyopathies, we also studied the organization of the intercalated disks, particularly the distribution of desmosomal proteins. Normal amounts of the major desmosomal proteins were found, but the intercalated disks were more convoluted and elongated and had a zigzag appearance. In this largest series to date of individuals with a single head domain DES mutation, patients show a variable yet predominantly cardiologic phenotype characterized by conduction disease at an early age and RV involvement including right bundle branch block and/or RV tachycardias and arrhythmogenic RV cardiomyopathy phenocopies. A localized effect of desmin on the structure of the cardiac intercalated disks might contribute to disease pathogenesis.