Merck
  • Detection of Pneumocystis infections by in situ hybridization in lung samples of Austrian pigs with interstitial pneumonia.
Detection of Pneumocystis infections by in situ hybridization in lung samples of Austrian pigs with interstitial pneumonia.
Medical mycology (2013-07-19)
Diana Binanti, Meike M Mostegl, Christiane Weissenbacher-Lang, Nora Nedorost, Herbert Weissenböck
ABSTRACT
Pneumocystis carinii f. sp. suis is a fungus multiplying in the respiratory tract of pigs which occasionally is associated with interstitial pneumonia. Identification of Pneumocystis in tissue samples is considered difficult and there are only scarce data on its occurrence in European pigs. This investigation presents an in situ hybridization (ISH) procedure for identification of Pneumocystis spp. in paraffin wax embedded tissue samples and its application for labeling the agent in lung samples of pigs with interstitial pneumonia. Thirty-two out of 100 lung samples from pigs on Austrian farms were identified as positive, five of them with multiple, 12 with moderate and 15 with few organisms but Grocott’s methenamine silver staining demonstrated that only 20 cases were unequivocally positive for Pneumocystis carinii. In addition to interstitial pneumonia Pneumocystis-positive pigs were more frequently affected with granulomatous pneumonia than Pneumocystis-negative pigs. Frequently concurrent infections with different viral or bacterial lung pathogens were noted but there was no positive correlation between Pneumocystis- and PCV-2-infections. With other infections, no clear-cut differences between Pneumocystis-positive and Pneumocystis-negative animals were found. This study shows that Pneumocystis infections occur frequently in Austrian pigs with interstitial pneumonia. It remains to be shown which are the factors triggering severe multiplication and whether infection with Pneumocystis alone is able to induce lung disease in pigs.
Sigma-Aldrich
Tetrazolium Blue Chloride, suitable for microbiology, ≥90% (T)
Sigma-Aldrich
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds