Merck
Synaptic transmission: two players team up for a new tune.
Current biology : CB (1998-11-21)
R Jahn
ABSTRACT
An unexpected link has been found between two molecules that were thought to perform seemingly unrelated functions. The AMPA-subclass glutamate receptor GluR2 interacts with the membrane-fusion protein NSF in a manner that appears important for receptor-mediated intracellular signalling.