Merck
Inorganic bismuth(III) compounds in the therapy.
Arzneimittel-Forschung (1978-01-01)
M Wesolowski
PMID350231
ABSTRACT
A survey is given of the pharmacopoeial preparations of inorganic Bi(III) compounds: bismuth subnitrate and bismuth subcarbonate. The use of these preparations, methods of preparation, qualitative and quantitative assays have been reviewed.