Merck
  • Contributions of BrCl, Br2, BrOCl, Br2O, and HOBr to regiospecific bromination rates of anisole and bromoanisoles in aqueous solution.
Contributions of BrCl, Br2, BrOCl, Br2O, and HOBr to regiospecific bromination rates of anisole and bromoanisoles in aqueous solution.
Environmental science & technology (2015-03-25)
John D Sivey, Mark A Bickley, Daniel A Victor
ABSTRACT
When bromide-containing waters are chlorinated, conventional wisdom typically assumes HOBr is the only active brominating agent. Several additional and often-overlooked brominating agents (including BrCl, Br2, BrOCl, Br2O) can form in chlorinated waters, albeit at generally lower concentrations than HOBr. The extent to which these additional brominating agents influence bromination rates of disinfection byproduct precursors is, however, poorly understood. Herein, the influence of BrCl, Br2, BrOCl, Br2O, and HOBr toward rates of sequential bromination of anisole was quantified. Conditions affecting bromine speciation (e.g., pH, concentrations of chloride, bromide, and chlorine) were varied, and regiospecific second-order rate constants were calculated for reactions of each brominating agent with anisole, 2-bromoanisole, and 4-bromoanisole. The regioselectivity of anisole bromination changed with pH, consistent with the participation of more than one brominating agent. Under conditions representative of chlorinated drinking water, contributions to bromination rates decreased as BrCl > BrOCl > HOBr > Br2O (Br2 negligible). The second-order rate constant determined for net bromination of anisole by HOBr is up to 3000-times less than reported in previous studies (which assumed HOBr was the only active brominating agent). Accordingly, models that assume HOBr is the only kinetically relevant brominating agent in solutions of free bromine may be insufficient for reactions involving modestly nucleophilic organic compounds.
Sigma-Aldrich
4-Bromoanisole, ≥99.0%