Merck
  • Trace analysis of biocidal oligoguanidines in environmental water samples.
Trace analysis of biocidal oligoguanidines in environmental water samples.
Journal of chromatography. A (2013-10-29)
Tamara Buchberger, Markus Himmelsbach, Wolfgang Buchberger
ABSTRACT
This paper demonstrates the determination of residues of biocidal oligoguanidines manufactured by polycondensation of guanidine hydrochloride and 2,2'-(ethylenedioxy)bis(ethylamine) in environmental water samples. The analytes were preconcentrated from samples adjusted to pH 4 by solid-phase extraction using a mixed-mode sorbent with weak cation exchange functionalities. Elution from the sorbent was achieved by 2M hydrochloric acid in methanol. After evaporation and reconstitution in water, the extract was analyzed by reversed-phase high performance liquid chromatography combined with quadrupole/time-of-flight or triple quadrupole mass spectrometry employing electrospray ionization. A preconcentration factor of 10,000 could be achieved and detection limits were in the sub-μgL(-1) range. This method looks promising to monitor the fate of these biocides released into the aquatic environment during different applications.
Sigma-Aldrich
2,2′-(Ethylenedioxy)bis(ethylamine), 98%