Merck
  • Chemical and biomimetic total syntheses of natural and engineered MCoTI cyclotides.
Chemical and biomimetic total syntheses of natural and engineered MCoTI cyclotides.
Organic & biomolecular chemistry (2008-04-04)
Panumart Thongyoo, Núria Roqué-Rosell, Robin J Leatherbarrow, Edward W Tate
ABSTRACT
The naturally-occurring cyclic cystine-knot microprotein trypsin inhibitors MCoTI-I and MCoTI-II have been synthesised using both thia-zip native chemical ligation and a biomimetic strategy featuring chemoenzymatic cyclisation by an immobilised protease. Engineered analogues have been produced containing a range of substitutions at the P1 position that redirect specificity towards alternative protease targets whilst retaining excellent to moderate affinity. Furthermore, we report an MCoTI analogue that is a selective low-microM inhibitor of foot-and-mouth-disease virus (FMDV) 3C protease, the first reported peptide-based inhibitor of this important viral enzyme.
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%