Merck
  • Synthesis and Spectroscopic Analysis of Novel 1H-Benzo[d]imidazoles Phenyl Sulfonylpiperazines.
Synthesis and Spectroscopic Analysis of Novel 1H-Benzo[d]imidazoles Phenyl Sulfonylpiperazines.
Pharmaceuticals (Basel, Switzerland) (2012-01-01)
Amjad M Qandil
ABSTRACT
A group of benzimidazole analogs of sildenafil, 3-benzimidazolyl-4-methoxy-phenylsulfonylpiperazines 2-4 and 3-benzimidazolyl-4-methoxy-N,N-dimethyl- benzenesulfonamide (5), were efficiently synthesized. Compounds 2-5 were characterized by NMR and MS and contrary to the reported mass spectra of sildenafil, the spectra of the piperazine-containing compounds 2-4 showed a novel fragmentation pattern leading to an m/z = 316. A mechanism for the formation of this fragment was proposed.
Sigma-Aldrich
1-Ethylpiperazine, 98%