Merck
Activation of a carbonyl compound by halogen bonding.
Chemical communications (Cambridge, England) (2014-05-07)
Stefan H Jungbauer, Sebastian M Walter, Severin Schindler, Laxmidhar Rout, Florian Kniep, Stefan M Huber
ABSTRACT
Using a prototypical Diels-Alder reaction as benchmark, we show that dicationic halogen-bond donors are capable of activating a neutral organic substrate. By various comparison experiments, the action of traces of acid or of other structural features of the halogen-bond donor not related to halogen bonding are excluded with high certainty.
Sigma-Aldrich
Schreiner′s Thiourea Catalyst, 95%