Merck
E/Z oxime isomerism in PhC(NOH)CN.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-02-13)
D Scott Bohle, Zhijie Chua, Inna Perepichka, Kristopher Rosadiuk
ABSTRACT
The reaction of nitric oxide with benzyl cyanide in the presence of potassium methoxide at low temperature gave the dipotassium salt of a bis-diazeniumdiolate 2 in excellent yield. Two new stereospecific syntheses of E or Z 2-(hydroxyimino)-2-phenylacetonitrile from 2 have been found. The thermodynamics of the E/Z isomerization has been investigated spectroscopically in solution, in the solid state by differential scanning calorimetry (DSC), and theoretically in the gas phase. Evidence of catalysis by NO of E/Z oxime isomerization has been observed.
Sigma-Aldrich
2-Hydroxyimino-2-phenylacetonitrile, mixture of syn and anti, 97%