Merck
  • Notch signaling induces cytoplasmic CD3 epsilon expression in human differentiating NK cells.
Notch signaling induces cytoplasmic CD3 epsilon expression in human differentiating NK cells.
Blood (2007-07-17)
Magda De Smedt, Tom Taghon, Inge Van de Walle, Greet De Smet, Georges Leclercq, Jean Plum
ABSTRACT
It has been proposed that heterogeneity in natural killer (NK)-cell phenotype and function can be achieved through distinct thymic and bone marrow pathways of NK-cell development. Here, we show a link between Notch signaling and the generation of intracellular CD3epsilon (cyCD3)-expressing NK cells, a cell population that can be detected in vivo. Differentiation of human CD34(+) cord blood progenitors in IL-15-supplemented fetal thymus organ culture or OP9-Delta-like 1 (DL1) coculture resulted in a high percentage of cyCD3(+) NK cells that was blocked by the gamma-secretase inhibitor DAPT. The requirement for Notch signaling to generate cyCD3(+) NK cells was further illustrated by transduction of CD34(+) cord blood (CB) cells with either the active intracellular part of Notch or the dominant-negative mutant of mastermind-like protein 1 that resulted in the generation of NK cells with respectively high or low frequencies of cyCD3. Human thymic CD34(+) progenitor cells displayed the potential to generate cyCD3(+) NK cells, even in the absence of Notch/DL1 signaling. Peripheral blood NK cells were unable to induce cyCD3 expression after DL1 exposure, indicating that Notch-dependent cyCD3 expression can only be achieved during the early phase of NK-cell differentiation.