Merck
  • Buccolam(®) (buccal midazolam): a review of its use for the treatment of prolonged acute convulsive seizures in the dental practice.
Buccolam(®) (buccal midazolam): a review of its use for the treatment of prolonged acute convulsive seizures in the dental practice.
British dental journal (2012-07-28)
P Jevon
ABSTRACT
This article is published in response to a query from reader Sarah Clements, based at Golding House Dental Practice Ltd in Kent, regarding the BDJ paper entitled 'Updated guidance on medical emergencies and resuscitation in the dental practice' (BDJ 2012; 212: 41-43). Sarah wished to be provided with further clarification on the ordering, prescribing and administration of midazolam.
Product Description
Supelco
Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®