- Ultralow doping in organic semiconductors: evidence of trap filling.
Ultralow doping in organic semiconductors: evidence of trap filling.
Physical review letters (2012-12-12)
Selina Olthof, Shafigh Mehraeen, Swagat K Mohapatra, Stephen Barlow, Veaceslav Coropceanu, Jean-Luc Brédas, Seth R Marder, Antoine Kahn
PMID23215211
ABSTRACT
Tail states in organic semiconductors have a significant influence on device performances by acting as traps in charge transport. We present a study of the controlled passivation of acceptor tail states in fullerene C(60) by the addition of electrons introduced by molecular n doping. Using ultralow doping, we are able to successively fill the traps with charges and examine the changes in conductivity, activation energy, mobility, and Fermi-level position. Passivation of the traps leads to an increase of the electron mobility in C(60) by more than 3 orders of magnitude, to reach 0.21 cm(2)/(V s).