Merck
Synthesis of new fused pyrimidines by isocyanate and isothiocyanate.
Chemical & pharmaceutical bulletin (2001-04-20)
A Z Chowdhury, Y Shibata
ABSTRACT
o-Aminonitrile or o-aminoester compounds were cyclized to fused pyrimidines by reacting with ethyl iso(thio)cyanatoacetate in pyridine, and then were methylated, halogenated and subsequently displaced by the amines studied.
Sigma-Aldrich
Ethyl isothiocyanatoacetate, 97%