- Synthesis and application of chiral N-heterocyclic carbene-oxazoline ligands: iridium-catalyzed enantioselective hydrogenation.
Synthesis and application of chiral N-heterocyclic carbene-oxazoline ligands: iridium-catalyzed enantioselective hydrogenation.
Two libraries of enantiomerically pure imidazolium salts bearing an oxazoline unit were synthesized. Deprotonation of the imidazolium salts and complexation of the resulting oxazoline-carbene ligands to iridium(I) was achieved in one step by mixing the imidazolium salts with NaOtBu and [(eta(4)-cod)IrCl](2) in THF at room temperature. The air-stable complexes were purified by flash chromatography. All complexes were analyzed by two-dimensional (2D) NMR methods and one compound from each family was characterized by X-ray structure analysis. The two libraries of iridium complexes were successfully tested in the asymmetric hydrogenation of unfunctionalized and functionalized olefins. Enantioselectivities of up to 90 % ee were obtained with trans-alpha-methylstilbene. Upon complexation of imidazolium salt 15 p with R(1) = phenyl, C-H bond activation of the phenyl ring gave rise to iridium(III) complex 17, which was fully characterized by NMR spectroscopy and X-ray structure analysis. Complex 17 proved to be catalytically inactive in the hydrogenation.