Merck
  • A computational approach to tuning the photochemistry of platinum(IV) anticancer agents.
A computational approach to tuning the photochemistry of platinum(IV) anticancer agents.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-07-19)
Hui-Chung Tai, Yao Zhao, Nicola J Farrer, Anna E Anastasi, Guy Clarkson, Peter J Sadler, Robert J Deeth
ABSTRACT
Diazido Pt(IV) complexes are inert stable prodrugs that can be photoactivated to produce Pt(II) species with promising anticancer activity. Our studies of the photochemistry of Pt(IV) complexes, [Pt(X)(2) (Y)(2) (Z)(2) ](0/-1) (X=N-ligands (NH(3) , pyridine, etc.)/S(CH(3) )(2) /H(-) , Y=(pseudo)halogen (N(3) (-) , I(-) ), Z=OR(-) , R=H, Ac) by time-dependent density functional theory (TDDFT) show close agreement with spectroscopic data. Broad exploration of cis/trans geometries, trans influences, the nature of the OR(-) and (pseudo)halogen ligands, electron-withdrawing/donating/delocalising substituents on the N-ligands, and intramolecular H bonds shows that: 1) the design of platinum(IV) complexes with intense bands shifted towards longer wavelengths (from 289 to ∼330 nm) can be achieved by introducing intramolecular H bonds involving the OH ligands and 2-hydroxyquinoline or by iodido ligands; 2) mesomeric electron-withdrawing substituents on pyridine result in low-energy absorption with significant intensity in the visible region; and 3) the distinct makeup of the molecular orbitals involved in the electronic transitions for cis/trans-{Pt(N(3) )(2) } isomers results in different photoproducts. In general, the comparison of the optimised geometries shows that Pt(IV) complexes with longer PtL bonds are more likely to undergo photoreduction with longer-wavelength light. The novel complex trans,trans,trans-[Pt(N(3) )(2) (OH)(2) (NH(3) )(4-nitropyridine)] with predicted absorption in the visible region has been synthesised. The experimental UV/Vis spectrum in aqueous solution correlates well with the intense band in the computed spectrum, whereas the overlay in the low-energy region can be improved by a solvent model. This combined computational and experimental study shows that TDDFT can be used to tune the coordination environment for optimising photoactive Pt(IV) compounds as anticancer agents.
Sigma-Aldrich
2-Hydroxyquinoline, 98%