Merck
  • Synthesis and biological evaluation of novel fumagillin and ovalicin analogues.
Synthesis and biological evaluation of novel fumagillin and ovalicin analogues.
Organic & biomolecular chemistry (2005-05-27)
Ralph Mazitschek, Axel Huwe, Athanassios Giannis
ABSTRACT
A promising approach among the numerous efforts to cure cancer is the interruption of the tumour-induced formation of new blood vessels (angiogenesis). By suppressing angiogenesis with drugs, the tumour can neither grow to a life threatening size, nor metastasize. The natural product fumagillin 1 and the structurally related ovalicin 2 are two of the most potent anti-angiogenic compounds. Here, we report the design and synthesis of novel fumagillin and ovalicin analogues lacking reactive epoxy functionalities, which were thought to be responsible for the severe toxic side-effects observed. We also report a new synthetic approach and the determination of the anti-angiogenic properties of these compounds in endothelial cells.
Sigma-Aldrich
Chloroacetyl isocyanate, ≥95%