Merck
  • Hydroxylation of the diterpenes ent-kaur-16-en-19-oic and ent-beyer-15-en-19-oic acids by the fungus Aspergillus niger.
Hydroxylation of the diterpenes ent-kaur-16-en-19-oic and ent-beyer-15-en-19-oic acids by the fungus Aspergillus niger.
Phytochemistry (2009-10-10)
Silvia Marquina, José Luis Parra, Manasés González, Alejandro Zamilpa, Jaime Escalante, María R Trejo-Hernández, Laura Alvarez
ABSTRACT
The diterpenes ent-kaur-16-en-19-oic acid (1) and ent-beyer-15-en-19-oic acid (2) are the major constituents of a spasmolytic diterpenic mixture obtained from the roots of Viguiera hypargyrea, a Mexican medicinal plant. Microbial transformation of 1 and 2 was performed with Aspergillus niger. Two metabolites, ent-7alpha,11beta-dihydroxy-kaur-16-en-19-oic acid (4) and ent-1beta,7alpha-dihydroxy-kaur-16-en-19-oic acid (5), were isolated from the incubation of 1, and one metabolite, ent-1beta,7alpha-dihydroxy-beyer-15-en-19-oic acid (6), was isolated in high yield (40%) from 2. The structures were elucidated on the basis of spectroscopic analyses and confirmed by X-ray crystallographic studies. Compounds 1-4 and 6 and methyl ester derivatives 4a and 6a were evaluated for their ability to inhibit the electrically induced contraction of guinea-pig ileum. Compounds 1, 3, 4, 4a and 5 were significantly active. These results showed that dihydroxylation of 1 at 7beta, 11alpha-, and 1alpha, 7beta-positions resulted in a loss of potency.
Sigma-Aldrich
Kaurenoic acid, ≥95% (LC/MS-ELSD)