Merck
  • The faciogenital dysplasia gene product FGD1 functions as a Cdc42Hs-specific guanine-nucleotide exchange factor.
The faciogenital dysplasia gene product FGD1 functions as a Cdc42Hs-specific guanine-nucleotide exchange factor.
The Journal of biological chemistry (1996-12-27)
Y Zheng, D J Fischer, M F Santos, G Tigyi, N G Pasteris, J L Gorski, Y Xu
ABSTRACT
The Rho family of small GTP-binding proteins plays important roles in the regulation of actin cytoskeleton organization and cell growth. Activation of these GTPases involves the replacement of bound GDP with GTP, a process catalyzed by the Dbl-like guanine-nucleotide exchange factors, all of which seem to share a putative catalytic motif termed the Dbl homology (DH) domain, followed by a pleckstrin homology (PH) domain. Here we have examined the role of a Dbl-like molecule, the faciogenital dysplasia gene product (FGD1), which when mutated in its Dbl homology domain, cosegregates with the developmental disease Aarskog-Scott syndrome. We report that a polypeptide of FGD1 encompassing the DH and PH domains can bind specifically to the Rho family GTPase Cdc42Hs and stimulates the GDP-GTP exchange of the isoprenylated form of Cdc42Hs. Microinjection of this FGD1 polypeptide into Swiss 3T3 fibroblast cells induces the formation of peripheral actin microspikes, similar to that previously observed when cells were injected with a constitutively active form of Cdc42Hs. This effect of FGD1 on actin organization is readily inhibited by coinjection of a dominant-negative mutant of Cdc42Hs. Examination of NIH 3T3 cells expressing the FGD1 fragment revealed that similar to cells expressing Dbl, two independent elements downstream of Cdc42Hs, the Jun NH2-terminal kinase and the p70 S6 kinase, became activated. Hence, our results indicate that FGD1, through its DH and PH domains, acts as a Cdc42Hs-specific guanine-nucleotide exchange factor and suggest that the Cdc42Hs GTPase may have a role in mammalian development.
Sigma-Aldrich
Anti-FGD1 antibody produced in rabbit, affinity isolated antibody