Merck
  • Histaminergic and serotonergic receptor blocking substances from the medicinal plant Garcinia mangostana.
Histaminergic and serotonergic receptor blocking substances from the medicinal plant Garcinia mangostana.
Planta medica (1996-10-01)
N Chairungsrilerd, K Furukawa, T Ohta, S Nozoe, Y Ohizumi
ABSTRACT
A crude methanolic extract of the fruit hull of Mangosteen, Garcinia mangostana L. inhibited the contractions of isolated thoracic rabbit aorta induced by histamine and serotonin. The extract of the fruit hull has been fractionated by silica gel chromatography, monitoring the pharmacological activity to give alpha- and gamma-mangostin. On the basis of pharmacological data, it is suggested that alpha-mangostin and gamma-mangostin are a histaminergic and a serotonergic receptor blocking agent, respectively.
Sigma-Aldrich
gamma-Mangostin, ≥98% (HPLC)