Merck
  • Multiple-dose pharmacokinetics of 12 milligrams of trimethoprim and 60 milligrams of sulfamethoxazole per kilogram of body weight per day in healthy volunteers.
Multiple-dose pharmacokinetics of 12 milligrams of trimethoprim and 60 milligrams of sulfamethoxazole per kilogram of body weight per day in healthy volunteers.
Antimicrobial agents and chemotherapy (1993-03-01)
R C Stevens, S C Laizure, P L Sanders, D S Stein
ABSTRACT
The disposition of 12 mg of trimethoprim and 60 mg of sulfamethoxazole per kg of body weight in six healthy male volunteers is described. The daily dose was evenly divided and administered orally every 6 h for 13 consecutive doses. Individual drug components were assayed by high-performance liquid chromatography. Steady-state concentrations in serum for trimethoprim and sulfamethoxazole were within the purported therapeutic ranges for treating Pneumocystis carinii pneumonia. Co-trimoxazole was well tolerated, and no subject withdrew from the study because of toxicity. Comparison of the pharmacokinetic parameters in this study with those of our previous findings indicates that the elimination of trimethoprim-sulfamethoxazole follows a first-order process within the dose ranges assessed. Administration of 15- to 20-mg/kg trimethoprim and 75- to 100-mg/kg sulfamethoxazole in four evenly divided doses for the first 24 h followed by 12 and 60 mg/kg/day, respectively, for the remainder of therapy rapidly attains concentrations in serum within the proposed P. carinii pneumonia therapeutic range. Clinical trials are indicated to evaluate this dosing scheme, which may decrease exposure to potentially excessive concentrations of trimethoprim and sulfamethoxazole.
Product Description
Sulfamethoxazole impurity A, European Pharmacopoeia (EP) Reference Standard