Merck
  • Activity of agricultural chemicals to modify mitomycin C induced growth inhibition of Bacillus subtilis in the rec assay.
Activity of agricultural chemicals to modify mitomycin C induced growth inhibition of Bacillus subtilis in the rec assay.
The Tohoku journal of experimental medicine (1988-08-01)
M Yamamoto, J Magara, S Takagi, N Shibuya, W S Chen, K Endoh
ABSTRACT
Rec assay of agricultural chemicals, MCPA-E, MCPA, CNP and PCNB was conducted. It was found that MCPA-E and MCPA accelerated the growth-inhibition originally induced by Mitomycin C in the rec- strain.
Product Description
Supelco
Chlornitrofen, PESTANAL®, analytical standard