Merck
  • Chitosan microparticles for sustaining the topical delivery of minoxidil sulphate.
Chitosan microparticles for sustaining the topical delivery of minoxidil sulphate.
Journal of microencapsulation (2011-08-10)
Guilherme Martins Gelfuso, Taís Gratieri, Patrícia Sper Simão, Luís Alexandre Pedro de Freitas, Renata Fonseca Vianna Lopez
ABSTRACT
Given the hypothesis that microparticles can penetrate the skin barrier along the transfollicular route, this work aimed to obtain and characterise chitosan microparticles loaded with minoxidil sulphate (MXS) and to study their ability to sustain the release of the drug, attempting a further application utilising them in a targeted delivery system for the topical treatment of alopecia. Chitosan microparticles, containing different proportions of MXS/polymer, were prepared by spray drying and were characterised by yield, encapsulation efficiency, size and morphology. Microparticles selected for further studies showed high encapsulation efficiency (∼82%), a mean diameter of 3.0 µm and a spherical morphology without porosities. When suspended in an ethanol/water solution, chitosan microparticles underwent instantaneous swelling, increasing their mean diameter by 90%. Release studies revealed that the chitosan microparticles were able to sustain about three times the release rate of MXS. This feature, combined with suitable size, confers to these microparticles the potential to target and improve topical therapy of alopecia with minoxidil.
Sigma-Aldrich
Minoxidil Sulfate