Merck
  • [The research practice of anti-arrhythmic agents targeting on potassium ion channel].
[The research practice of anti-arrhythmic agents targeting on potassium ion channel].
Yao xue xue bao = Acta pharmaceutica Sinica (2011-04-07)
Qian Yang, Xiao-Jian Wang, Yi-Qun Tang, Qi-Dong You
ABSTRACT
Due to the complicated pathogenesis of cardiac arrhythmia, the safe and effective therapeutic strategies for cardiac arrhythmia remain an urgent medical problems in the recent years. In this paper, we introduced the research practice of anti-arrhythmic agents targeting on potassium ion channel. The research progress of anti-arrhythmic agents in up-to-date literatures were also reviewed and prospected.
Sigma-Aldrich
Azimilide dihydrochloride, ≥97% (HPLC)