Merck
  • Removal of the toxic effects of chlormadinon acetate on the development of Drosophila melanogaster via the use of nordihydroguaiaretic acid.
Removal of the toxic effects of chlormadinon acetate on the development of Drosophila melanogaster via the use of nordihydroguaiaretic acid.
Toxicology and industrial health (2010-08-18)
D Altun, H Uysal, A Ayar, H Askin
ABSTRACT
In this study, the effects of chlormadinon acetate (CMA) and CMA + nordihydroguaiaretic acid (NDGA) on various developmental stages of Drosophila melanogaster were investigated. Different concentrations of CMA (1.0; 3.0; 5.0 and 10.0 μM/100 mL medium) and CMA + NDGA as the concentrations of CMA (1.0 + 1.0; 3.0 + 3.0; 5.0 + 5.0 and 10.0 + 10.0 μM/100 mL medium) were carried out during the developmental periods of the flies. When F(1) progeny of control and application groups were compared, CMA was found to extend the process of metamorphosis and decrease the total offspring numbers. However, these negative effects were inhibited by NDGA treatment at different concentrations. These results suggest that NDGA could effectively inhibit CMA-induced abnormalities in developmental stages of D. melanogaster. It was found that the difference between the groups was significantly important (p < 0.05).
Sigma-Aldrich
Chlormadinone acetate, ≥98%