Merck
  • [The effect of diabeton and metformin on the free-radical oxidation processes and the structural characteristics of the phospholipid composition of the erythrocyte membranes in non-insulin-dependent diabetes mellitus].
[The effect of diabeton and metformin on the free-radical oxidation processes and the structural characteristics of the phospholipid composition of the erythrocyte membranes in non-insulin-dependent diabetes mellitus].
Sovetskaia meditsina (1991-01-01)
T A Buĭdina, G S Kozlov, M I Balabolkin
ABSTRACT
Newly detected noninsulin--dependent diabetes mellitus is shown to have changes in phospholipid structure of erythrocytic membranes: a fall in total phospholipids, sphingomyelin, phosphatidylcholine, phosphatidic acid, a trend to increasing lysophosphatidylcholine. The shifts go in parallel with lipid peroxidation activation. Carbohydrate metabolism compensation is not enough to eradicate the shifts. The process is also dependent on the properties of a sugar-reducing drug. Diabeton is recommended as beneficial in this respect, while metformin is efficient in combinations.
Sigma-Aldrich
2-Amino-5-hydroxybenzoic acid, 98%