Merck
  • alpha-Isopropylmalate, a leucine biosynthesis intermediate in yeast, is transported by the mitochondrial oxalacetate carrier.
alpha-Isopropylmalate, a leucine biosynthesis intermediate in yeast, is transported by the mitochondrial oxalacetate carrier.
The Journal of biological chemistry (2008-08-07)
Carlo M T Marobbio, Giulia Giannuzzi, Eleonora Paradies, Ciro L Pierri, Ferdinando Palmieri
ABSTRACT
In Saccharomyces cerevisiae, alpha-isopropylmalate (alpha-IPM), which is produced in mitochondria, must be exported to the cytosol where it is required for leucine biosynthesis. Recombinant and reconstituted mitochondrial oxalacetate carrier (Oac1p) efficiently transported alpha-IPM in addition to its known substrates oxalacetate, sulfate, and malonate and in contrast to other di- and tricarboxylate transporters as well as the previously proposed alpha-IPM transporter. Transport was saturable with a half-saturation constant of 75 +/- 4 microm for alpha-IPM and 0.31 +/- 0.04 mm for beta-IPM and was inhibited by the substrates of Oac1p. Though not transported, alpha-ketoisocaproate, the immediate precursor of leucine in the biosynthetic pathway, inhibited Oac1p activity competitively. In contrast, leucine, alpha-ketoisovalerate, valine, and isoleucine neither inhibited nor were transported by Oac1p. Consistent with the function of Oac1p as an alpha-IPM transporter, cells lacking the gene for this carrier required leucine for optimal growth on fermentable carbon sources. Single deletions of other mitochondrial carrier genes or of LEU4, which is the only other enzyme that can provide the cytosol with alpha-IPM (in addition to Oac1p) exhibited no growth defect, whereas the double mutant DeltaOAC1DeltaLEU4 did not grow at all on fermentable substrates in the absence of leucine. The lack of growth of DeltaOAC1DeltaLEU4 cells was partially restored by adding the leucine biosynthetic cytosolic intermediates alpha-ketoisocaproate and alpha-IPM to these cells as well as by complementing them with one of the two unknown human mitochondrial carriers SLC25A34 and SLC25A35. Oac1p is important for leucine biosynthesis on fermentable carbon sources catalyzing the export of alpha-IPM, probably in exchange for oxalacetate.
Sigma-Aldrich
2-Isopropylmalic acid, 98%