Merck
  • Urine concentrations of fentanyl and norfentanyl during application of Duragesic transdermal patches.
Urine concentrations of fentanyl and norfentanyl during application of Duragesic transdermal patches.
Journal of analytical toxicology (2004-11-02)
Alphonse Poklis, Ronald Backer
ABSTRACT
A study of the urinary concentration of fentanyl (F) and its major metabolite norfentanyl (NF) in chronic pain patients treated with the Duragesic continuous release transdermal patches is presented. These patches are available in 10, 20, 30, and 40 cm(2) sizes releasing 25, 50, 75, and 100 microg/h F, respectively. F is rapidly and extensively metabolized, with NF as the major metabolite. Five hundred-forty six random urine specimens were collected from chronic pain patients wearing 25, 50, 75, or 100 ug F transdermal patches. Urine specimens were collected from hours after application to several days later after continuous F release. Each specimen was analyzed for F, NF, creatinine, and pH. Additionally, each was screened by enzyme immunoassay for the following: amphetamines, barbiturates, benzodiazepines, cocaine metabolite, methadone, phencyclidine, d-propoxyphene, opiates, and marijuana metabolites. All positive screening results were confirmed by gas chromatography-mass spectrometry (GC-MS). F and NF were isolated from urine by solid-phase extraction then identified and quantified by GC-MS in SIM mode. The LODs and LOQs for F and NF were 3 ng/mL, respectively. The results of F and NF analysis of urine form those wearing 25-microg patches (N = 142) was mean F, 47 ng/mL with a range of 0 to 983 ng/mL, and 97% of the specimens contained < 200 ng/mL and mean NF, 175 ng/mL with a range of 0-980 ng/mL, while 95% of the specimens contained < 400 ng/mL. The results of F and NF analysis of urine form those wearing 50 microg patches (N = 184) was: mean F, 74 ng/mL with a range of 0 to 589 ng/mL, and 92% of the specimens contained < 200 ng/mL and mean NF, 257 ng/mL with a range of 0-2200 ng/mL, and 98% of the specimens contained < 1000 ng/mL. The results of F and NF analysis of urine form those wearing 75 microg patches (N = 85) was mean F, 107 ng/mL with a range of 0 to 1280 ng/mL, and 98% of the specimens contained < 400 ng/mL and mean NF, 328 ng/mL with a range of 0-5630 ng/mL, and 99% of the specimens contained < 1000 ng/mL. The results of F and NF analysis of urine form those wearing 100 ug patches (N = 135) was mean F, 100 ng/mL with a range of 0 to 1080 ng/mL, while 96% of the specimens contained < 400 ng/mL and mean NF, 373 ng/mL with a range of 0-5730 ng/mL, and 95% of the specimens contained < 1000 ng/mL. The incidence of other drugs detected as a percentage the specimens was opiates, 48%, benzodiazepines, 43%; barbiturates, 3%; methadone, 4%; marijuana metabolite, 3%; and cocaine metabolite, 1%. With the exception of F and/or NF, no other drugs were detected in 25% of the specimens. These data demonstrate the wide variation in concentrations of F and NF in random urine specimens following application of Duragesic patches. However, these values obtained during therapeutic use far exceed concentrations previously reported in fatal poisoning. In general, one may expect to find urine NF concentrations 3-4 times higher than those of F.
Product Description
Supelco
Norfentanyl oxalate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®