Merck
A simple fluorometric determination of vitamin C.
Chemical & pharmaceutical bulletin (1998-10-17)
K Mori, M Kidawara, M Iseki, C Umegaki, T Kishi
ABSTRACT
A simple and accurate method for determination of vitamin C (ascorbic acid (AsA) and dehydroascorbic acid (DHA)) using 4,5-dimethyl-o-phenylenediamine (DMPD) was investigated. It was found that DMPD is a useful fluorescent reagent. The reaction product of DMPD with DHA showed strong and stable fluorescence (Ex; 360 nm, Em; 440 nm). Fluorometric derivatives were extracted with isobutanol or n-butanol. Extraction with isobutanol was superior to that with n-butanol in terms of specificity, since fluorometric derivatives of keto acids were extracted with n-butanol, together with DHA. The fluorescence intensity of DMPD derivatives was absolutely stable in isobutanol for at least 24 h. The sensitivity of determination of vitamin C was improved by removing several non-fluorometric compounds coexisting in the samples. The derivative derived from AsA was easily separated from those of keto acids by an HPLC method. The determination of vitamin C in natural products was thus improved by extraction and the HPLC method.
Sigma-Aldrich
4,5-Dimethyl-1,2-phenylenediamine, 98%