Merck
  • Considerations on the new nevirapine: switching patients from twice-daily to once-daily.
Considerations on the new nevirapine: switching patients from twice-daily to once-daily.
Journal of the International Association of Providers of AIDS Care (2013-03-02)
Douglas Ward, Jihad Slim
ABSTRACT
Nevirapine (NVP) was the first nonnucleoside reverse transcriptase inhibitor (NNRTI) approved for the treatment of HIV infection. NVP can provide safe and efficacious viral suppression for treatment-naive patients and for virologically controlled patients "switching" from other NNRTI or protease inhibitor-based regimens. Formulations allowing once-daily dosing of antiretrovirals can significantly improve regimen adherence, which is important for maintaining virologic control, especially for NNRTI-based regimens with low barriers for genetic resistance. Randomized and controlled clinical trials have established the clinical non inferiority of a new, extended-release formulation (XR) of NVP, in both treatment-naive (VERxVE) and treatment-experienced patients (TRANxITION), where patients already stable on the immediate-release formulation of NVP were safely transitioned directly to NVP XR. As a potentially more convenient once-daily option, NVP XR may improve adherence and reduce the risk of mutant viruses, attendant virologic failure, and the spread of drug resistance.
Sigma-Aldrich
Nevirapine