Merck
  • Demethylwedelolactone derivatives inhibit invasive growth in vitro and lung metastasis of MDA-MB-231 breast cancer cells in nude mice.
Demethylwedelolactone derivatives inhibit invasive growth in vitro and lung metastasis of MDA-MB-231 breast cancer cells in nude mice.
European journal of medicinal chemistry (2012-08-29)
Yean-Jang Lee, Wea-Lung Lin, Nai-Fang Chen, Shien-Kai Chuang, Tsui-Hwa Tseng
ABSTRACT
The anticancer properties of demethylwedelolactone (DWEL) and wedelolactone (WEL), which are naturally occurring coumestans, have not been well characterized. In this study, we investigated the anti-invasive effects of synthetic WEL and DWEL on human MDA-MB-231 breast cancer cells. We found that WEL and DWEL inhibited the anchorage-independent growth and also suppressed cell motility and cell invasion of MDA-MB-231 cells. In addition, WEL and DWEL reduced the activity and expression of matrix metalloproteinases (MMPs) involved in blocking the IκB-α/NFκB and MEK/ERK signaling pathways in MDA-MB-231 cells. Furthermore, DWEL suppressed the metastasis and lung colonization of the tumor cells in the nude mice. Altogether, these data suggest that DWEL derivatives exert anti-invasive growth effect on breast cancer cells.
Sigma-Aldrich
Demethylwedelolactone, ≥98% (HPLC)
Sigma-Aldrich
Wedelolactone, ≥98% (HPLC), powder