Merck
Altered ion-responsive gene expression in Mmp20 null mice.
Journal of dental research (2010-10-12)
C E Tye, R Sharma, C E Smith, J D Bartlett
ABSTRACT
During enamel maturation, hydroxyapatite crystallites expand in volume, releasing protons that acidify the developing enamel. This acidity is neutralized by the buffering activity of carbonic anhydrases and ion transporters. Less hydroxyapatite forms in matrix metalloproteinase-20 null (Mmp20(-/-)) mouse incisors, because enamel thickness is reduced by approximately 50%. We therefore asked if ion regulation was altered in Mmp20(-/-) mouse enamel. Staining of wild-type and Mmp20(-/-) incisors with pH indicators demonstrated that wild-type mice had pronounced changes in enamel pH as development progressed. These pH changes were greatly attenuated in Mmp20(-/-) mice. Expression of 4 ion-regulatory genes (Atp2b4, Slc4a2, Car6, Cftr) was significantly decreased in enamel organs from Mmp20(-/-) mice. Notably, expression of secreted carbonic anhydrase (Car6) was reduced to almost undetectable levels in the null enamel organ. In contrast, Odam and Klk4 expression was unaffected. We concluded that a feedback mechanism regulates ion-responsive gene expression during enamel development.
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %
Millipore
Methyl Red solution, suitable for microbiology