Merck
  • Syntheses, characterization and biological activities of rare earth metal complexes with curcumin and 1,10-phenanthroline-5,6-dione.
Syntheses, characterization and biological activities of rare earth metal complexes with curcumin and 1,10-phenanthroline-5,6-dione.
Journal of inorganic biochemistry (2009-01-13)
Yu-Min Song, Jun-Peng Xu, Lan Ding, Qian Hou, Jing-Wang Liu, Zao-Long Zhu
ABSTRACT
Three new solid complexes have been synthesized by the reaction of rare earth(III) nitrate with the first ligand curcumin (HL) and the second ligand 1,10-phenanthroline-5,6-dione (L') in alcohol solution (pH=6.5-7.0). The composition of the complexes has been characterized by elemental analysis, molar conductivity, thermogravimetric analysis, IR, UV-vis methods. The results reveal that beta-diketone group of the first ligand to coordinates with rare earth ions in bidentate mode after deprotonated. But the second ligand uses its two N atoms coordinates with rare earth ions in bidentate mode. The general formula of the complexes is REL(3)L' (RE=Sm, Eu, Dy). The results of antibacterial activity indicated that the complexes have excellent antibacterial ability for the testing bacterium than that of curcumin. The result of agarose gel electrophoresis suggested that the complex of SmL(3)L' can cleave the plasmid DNA at physiological pH and temperature. And it was found that the cleavage process of plasmid DNA was sensitive to pH, however, adding radical scavengers almost had no effect on the DNA cleavage reaction, therefore, the cleavage of DNA by SmL(3)L' does not produce diffusible hydroxyl radicals via the Fenton reaction.
Sigma-Aldrich
1,10-Phenanthroline-5,6-dione, 97%