Merck
  • Selectivity of inhibitors of endocannabinoid biosynthesis evaluated by activity-based protein profiling.
Selectivity of inhibitors of endocannabinoid biosynthesis evaluated by activity-based protein profiling.
Bioorganic & medicinal chemistry letters (2008-07-29)
Heather S Hoover, Jacqueline L Blankman, Sherry Niessen, Benjamin F Cravatt
ABSTRACT
The endocannabinoid 2-arachidonoylglycerol (2-AG) has been implicated as a key retrograde mediator in the nervous system based on pharmacological studies using inhibitors of the 2-AG biosynthetic enzymes diacyglycerol lipase alpha and beta (DAGL-alpha/beta). Here, we show by competitive activity-based protein profiling that the DAGL-alpha/beta inhibitors, tetrahydrolipstatin (THL) and RHC80267, block several brain serine hydrolases with potencies equal to or greater than their inhibitory activity against DAGL enzymes. Interestingly, a minimal overlap in target profiles was observed for THL and RHC80267, suggesting that pharmacological effects observed with both agents may be viewed as good initial evidence for DAGL-dependent events.
Sigma-Aldrich
RHC 80267, ≥98% (HPLC), solid