Merck
  • Evolved CYP102A1 (P450BM3) variants oxidise a range of non-natural substrates and offer new selectivity options.
Evolved CYP102A1 (P450BM3) variants oxidise a range of non-natural substrates and offer new selectivity options.
Chemical communications (Cambridge, England) (2008-02-20)
Christopher J C Whitehouse, Stephen G Bell, Henry G Tufton, Richard J P Kenny, Lydia C I Ogilvie, Luet-Lok Wong
ABSTRACT
The evolution of CYP102A1 variants with enhanced activity and altered specificity characteristics.
Sigma-Aldrich
Propylbenzene, ≥99.0% (GC)
Sigma-Aldrich
Propylbenzene, 98%
Supelco
Propylbenzene, analytical standard