Merck
  • Surface-enhanced Raman spectroscopy for uranium detection and analysis in environmental samples.
Surface-enhanced Raman spectroscopy for uranium detection and analysis in environmental samples.
Analytica chimica acta (2007-11-21)
Chuanmin Ruan, Wensui Luo, Wei Wang, Baohua Gu
ABSTRACT
Techniques for rapid screening of uranium in environmental samples are needed, and this study entails the development of surface-enhanced Raman scattering (SERS) for analyzing uranium in aqueous media with improved sensitivity and reproducibility. A new SERS substrate based on (aminomethyl)phosphonic acid (APA)-modified gold nanoparticles was found to give greater than three orders of magnitude SERS enhancement compared with unmodified bare gold nanoparticles. Intensities of uranyl band at about 830 cm(-1) were proportional to the concentrations of uranium in solution, especially at relatively low concentrations (<10(-5) M). A detection limit of approximately 8x10(-7) M was achieved with a good reproducibility since the measurement was performed directly in dispersed aqueous suspension. Without pretreatment, the technique was successfully employed for detecting uranium in a highly contaminated groundwater with a low pH, high dissolved salts (e.g., nitrate, sulfate, calcium and aluminum) and total organic carbon.
Sigma-Aldrich
(Aminomethyl)phosphonic acid, 99%