Merck
  • Monitoring paracetamol metabolism after single and repeated administration in pediatric patients with neoplastic diseases.
Monitoring paracetamol metabolism after single and repeated administration in pediatric patients with neoplastic diseases.
International journal of clinical pharmacology and therapeutics (2007-10-03)
S Koling, G Hempel, C Lanvers, J Boos, G Würthwein
ABSTRACT
Paracetamol (PCM) is frequently used in pediatric patients with neoplastic disease. It is metabolized mainly by conjugation, but at therapeutic concentrations, a small fraction of the drug undergoes oxidative metabolism via cytochrome P450 forming the hepatotoxic intermediate N-acetyl-p-benzo-quinone-imine (NAPQI) which is usually conjugated with glutathione and excreted as paracetamol mercapturate and paracetamol cysteine. The aim of this monitoring study was to evaluate PCM metabolism with minimal intervention during routine treatment with single and repeated administration in patients undergoing antineoplastic therapy. A total of 107 urine samples collected 4-12 h after PCM administration from 29 children undergoing antineoplastic treatment, and 10 children without antineoplastic treatment were analyzed for PCM, PCM glucuronide (PCM-G), PCM sulfate (PCM-S), PCM mercapturate (PCM-M) and PCM cysteine (PCM-C). The median (range) percentages for metabolites in urine were: a) in children with and without chemotherapy after the first administration: PCM: 0 (0-100) and 4 (0-11)%, PCM-G: 55 (0-88) and 51 (18 - 68)%, PCM-S: 30 (0-73) and 32 (22-57)%, PCM-(M+C): 13 (0-52) and 9 (0-24)%, respectively; b) after repeated administration in children with chemotherapy: PCM: 0 (0-51)%, PCM-G: 42 (7-100)%, PCM-S: 28 (0-70)%, PCM-(M+C): 24 (0-66)%. The pattern of PCM excretion in children undergoing antineoplastic treatment regimens is highly variable. Repeated administration is associated with a significant increase in the products of oxidative metabolism. This might indicate an increase in metabolism via the hepatotoxic NAPQI.
Sigma-Aldrich
Paracetamol sulfate potassium salt, solid, ≥97% (HPLC)
Sigma-Aldrich
N-Acetylbenzoquinoneimine
Supelco
Paracetamol β-D-glucuronide, analytical standard