Merck
  • LTA-mediated synthesis and complete assignment of (1)H and (13)C NMR data of two natural 11-nordrimanes: isonordrimenone and polygonone.
LTA-mediated synthesis and complete assignment of (1)H and (13)C NMR data of two natural 11-nordrimanes: isonordrimenone and polygonone.
Magnetic resonance in chemistry : MRC (2007-09-19)
Luis Moreno-Osorio, Luis Espinoza, Mauricio Cuellar, Marcelo Preite
ABSTRACT
Two naturally occurring 11-nordrimanes were synthesized, and their (1)H and (13)C NMR spectra were unambiguously assigned in full for the first time.