Merck
  • Mutational analysis of the active center of plant fructosyltransferases: Festuca 1-SST and barley 6-SFT.
Mutational analysis of the active center of plant fructosyltransferases: Festuca 1-SST and barley 6-SFT.
FEBS letters (2005-08-16)
Denise Altenbach, Eveline Nüesch, Tita Ritsema, Thomas Boller, Andres Wiemken
ABSTRACT
The active center of the glycoside hydrolase family 32 contains the three characteristic motifs (N/S)DPNG, RDP, and EC. We replaced the N-terminal region including the (N/S)DPNG motif of barley 6-SFT (sucrose:fructan 6-fructosyltransferase) by the corresponding region of Festuca 1-SST (sucrose:sucrose 1-fructosyltransferase). The chimeric enzyme, expressed in Pichia, retained the specificity of 6-SFT. Attempts to replace a larger piece at the N-terminus including also the RDP motif failed. A point mutation introduced in the RDP motif of 1-SST abolished enzymatic activity. Interestingly, point mutations of the EC-motif resulted in an enzyme which had lost the capability to form 1-kestose and glucose from sucrose but still accepted 1-kestose, producing fructose and sucrose as well as nystose.
Product Description
Supelco
1-Kestose, analytical standard