- [Inhibitory effect of zinc on beta-hexosaminidase release from RBL-2H3 cells by synthetic chemicals].
[Inhibitory effect of zinc on beta-hexosaminidase release from RBL-2H3 cells by synthetic chemicals].
The effects of foods and chemicals related to food hygiene on degranulation were evaluated using a method for assaying the enzyme activity of beta-hexosaminidase as an index of chemical mediator release from RBL-2H3 cells in vitro. Using a previously developed assay system, we had found a large number of inhibitors and promoters of degranulation of RBL-2H3 cells. In the present study, we examined the inhibitory effect of zinc chloride on the degranulation (beta-hexosaminidase release) from RBL-2H3 cells with or without antigen in the presence of the degranulation-promotive chemicals, namely, 4 food additives, 7 pesticides and 2 veterinary drugs. These promotive chemicals were classified into two types on the basis of inhibitory profile by zinc chloride: 1) those which showed marked degranulation-inhibitory action when the cells were stimulated with antigen, such as butylhydroxyanisole, dibutylhydroxytoluene, EPN, cis- and trans-permethrin, prothiofos, pyridaben, terbufos, 2) those which showed marked degranulation-inhibitory action whether the cells were stimulated with antigen or not, such as butyl p-hydroxybenzoate, o-phenylphenol, bitertanol, salinomycin. In conclusion, zinc had a dramatic inhibitory effect on enhanced degranulation induced by synthetic chemicals in vitro.