Merck
  • alpha,alpha-gem-Difluorination of alpha-(alkylthio)acetophenone derivatives with N-fluoropyridinium salts.
alpha,alpha-gem-Difluorination of alpha-(alkylthio)acetophenone derivatives with N-fluoropyridinium salts.
Chemical & pharmaceutical bulletin (2000-08-03)
S Takeda, Y Kaneko, H Eto, M Tokizawa, S Sato, K Yoshida, S Namiki, M Ogawa
ABSTRACT
The alpha,alpha-gem-difluorination of 2',4'-difluoro-alpha-(methylthio)acetophenone (1a) with N-fluoropyridinium salts gave 2',4',alpha,alpha-tetrafluoro-alpha-(methylthio)acetophenone (3a). This reaction was accelerated by the addition of zinc chloride, zinc bromide or anhydrous iron(III) chloride, and higher yields than the reaction without additives were obtained. The gem-difluorination reaction using FP-T300 in the presence of zinc bromide was applicable to other alpha-(alkylthio)acetophenone derivatives (1).
Sigma-Aldrich
Zinc bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, 99.999% trace metals basis
Sigma-Aldrich
Zinc bromide, puriss., anhydrous, ≥98%