Merck
  • Lack of a significant effect of arctiin on development of 7,12-dimethylbenz(a)anthracene-induced mammary tumors in ovariectomized Sprague-Dawley rats.
Lack of a significant effect of arctiin on development of 7,12-dimethylbenz(a)anthracene-induced mammary tumors in ovariectomized Sprague-Dawley rats.
Nutrition and cancer (2007-06-19)
Mai Hasumura, Makoto Ueda, Jun-ichi Onose, Toshio Imai, Masao Hirose
ABSTRACT
Arctiin, a plant lignan, is metabolized to hormone-like compounds with weak estrogenic and antioxidative activity in experimental animals and man. To clarify its influence on mammary carcinogenesis, female rats were administrated 7,12-dimethylbenz(a)anthracene (DMBA) once, and when the incidence of palpable mammary tumors reached 50%, subjected to ovariectomy (OVX) and divided into tumor-bearing [DMBA-Tumor (+)] and no-tumor-bearing [DMBA-Tumor (-)] groups, subgroups of each then being fed soybean-free diet containing 0, 40, 200, and 1000 ppm of arctiin for 31 wk. The incidence and multiplicity of palpable tumors in the 200 ppm DMBA-Tumor (+) subgroup from week 12 of arctiin treatment tended to be decreased as compared to the 0 ppm subgroup and at terminal sacrifice, the volume of histopathologically defined mammary tumors was decreased in the 40 ppm DMBA-Tumor (-) subgroup, but again without statistical significance. In conclusion, weak inhibitory effects of arctiin on DMBA-induced mammary tumor development were suggested in OVX rats, but any further assessment is needed to obtain conclusive results.
Sigma-Aldrich
Arctiin, ≥95% (LC/MS-ELSD)