Merck
Butvar B-98 as a thin support film.
Ultramicroscopy (1979-01-01)
D A Handley, B R Olsen
PMID531984
ABSTRACT
Support films prepared from butvar B-98 resin are mechanically stable, electron transparent, and possess minimum intrinsic structure. A simple procedure for routine preparation of support films using this resin is provided.